missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Nylon Biopsy Bags
Ensure unrestricted fluid movement around tissue with Epredia™ Nylon Biopsy Bags.
Brand: Epredia™ 6774009
346.20 EUR valid until 2024-12-31
Use promo code "21628" to get your promotional price.
This item is not returnable.
View return policy
Description
- Reduce risk of small specimen loss
- Reduce reagent carryover and pressure artifact often found with sponges
- Remove specimen rapidly with convenient peel-open bag
Dimensions:
- Small Biopsy Bags measure 1.2 × 1.7″ (31 × 43mm)
- Medium Biopsy Bags measure 1.8 × 2.6″ (45 × 65mm)
- Large Biopsy Bags measure 3 × 3.5″ (75 × 85mm)
Specifications
Biopsy Bag | |
Blue | |
65 mm | |
2.6 in. | |
1000/Pk. |
Nylon | |
45 mm | |
1.8 in. | |
Medium |