Learn More
Masterflex™ Nylon Female Luer Fitting
Use these low-profile hose barbs with semi-rigid tubing such as nylon, polyethylene and polypropylene. Nylon Female Luer Fittings feature a smooth bore hose barb to reduce flow turbulence and risk of line contamination.
Brand:  Masterflex™ 45502-02
 View more versions of this product
Product Code. 13525770
  • 17.55 € / Pack of 25
Estimated Shipment: 14-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Standard hose barb for use with flexible tubing
  • Smooth bore hose barb to reduce flow turbulence and risk of line contamination
  • Wide-bore hose barb for enhanced fluid path flow
Specifications
Specifications
Female Luer Fitting
Nylon
Fits Female luer x 3/32 in. hose barb adapter
25 Pack