Learn More
Cole-Parmer™ Essentials Disposable Forceps, Reinforced Nylon, Serrated Pointed Tips
Get a more precise grip with these Reinforced Nylon Disposable Forceps with Serrated Pointed Tips.
Brand: Cole-Parmer™ Essentials 06443-25
This item is not returnable.
View return policy
Description
- Corrugated handles provide you with a secure grip.
- Reduce cross contamination with these disposable forceps.
- Disposable.
- Forceps are available in Polystyrene and reinforced Nylon.
- Choose from sterile, autoclavable, as well as blunt or pointed tips.
- Sterile forceps are individually wrapped with easy to open peel-packs.
- Non-sterile forceps are autoclavable.
- Serrated pointed tips are ideal for handling smaller objects.
- Serrated blunt tips are more for general use.
Specifications
Pointed | |
4.5 in. | |
114 mm | |
100 Pk. |
100 Pk. | |
Nylon | |
Forceps | |
Forceps |
Product Content Correction
Product Title
Cole-Parmer™ Essentials Disposable Forceps, Reinforced Nylon, Serrated Pointed Tips > Disposable Forceps; Length: 114.3 mm (4.5 in.)
Spot an opportunity for improvement?Share a Content Correction