missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Nylon Large Lockout Pouch

Create your own custom lockout kit and use the pouch to store it neatly. Nylon Large Lockout Pouch comes with an attachment loop on the back.

Brand:  Brady™ 050979

Product Code. 11711865

  • 71.00 € / Each
Estimated Shipment: 09-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Contents not included.
  • Dimensions: 76.2 x 177.8 x 203.2 mm (D x W x H)
Specifications

Specifications

Lockout Belt Pouch
White on Red
1 Ea.
LARGE NYLON LOCKOUT POUCH W/BRADY LOGO
Large Lockout Pouch
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Brady™ Nylon Large Lockout Pouch > Large Lockout Pouch; Dimensions (D x W x H): 76 x 178 x 203 mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.