missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Nylon Membrane Filter
Naturally hydrophilic membrane designed to wet out evenly and retain its superior strength during use in general filtration or medical assays.
Brand: Fisherbrand™ 0462A00023
64.80 EUR valid until 2024-12-31
Use promo code "21931" to get your promotional price.
Description
- Naturally hydrophilic membrane
- Designed to wet out evenly and retain its superior strength during use in general filtration or medical assays
- Flexible, durable and tear resistant
- Autoclaved at 135°C
- Effective chemical compatibility with aqueous and alcoholic solutions and solvents
Specifications
Autoclavable | |
White | |
Nylon | |
Non-sterile | |
Sterilizing filtration of aqueous solutions, organic solvents, Bacterial and particles removal (blood Glucose, Serum Cholesterol, drugs) | |
Hydrophilic | |
Plain Disc | |
47 mm |
3.4bar | |
Microfilter | |
0.22 μm | |
9.9mL/min./cm2 | |
1.7L/min./cm2 | |
100 Pack | |
Yes |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction