missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Nylon Membrane Filter

Naturally hydrophilic membrane designed to wet out evenly and retain its superior strength during use in general filtration or medical assays.

Brand:  Fisherbrand™ 0464A00022

36.87 EUR valid until 2024-12-31
Use promo code "21931" to get your promotional price.


 View more versions of this product

Product Code. 11755315

  • 45.40 € / Pack of 100
Estimated Shipment: 11-12-2024
to see stock.
Explore more special offers
Add to basket

Description

Description

  • Naturally hydrophilic membrane
  • Designed to wet out evenly and retain its superior strength during use in general filtration or medical assays
  • Flexible, durable and tear resistant
  • Autoclaved at 135°C
  • Effective chemical compatibility with aqueous and alcoholic solutions and solvents
Specifications

Specifications

Autoclavable
White
Nylon
Non-sterile
Sterilizing filtration of aqueous solutions, organic solvents, Bacterial and particles removal (blood Glucose, Serum Cholesterol, drugs)
Hydrophilic
Plain Disc
25 mm
2bar
Microfilter
0.45 μm
26.9mL/min./cm2
3.2L/min./cm2
100 Pack
Yes
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers