Learn More
Brady™ Nylon Single Pole Circuit Breaker Lockout
Nylon Single Pole Circuit Breaker Lockout designed for 120 Volt breakers that have holes in the switch tongue. Quick and easy - just snap in place and apply lock. Dual cavities accommodate different size switch tongues.
Brand:  Brady™ 065387
Product Code. 11741635
  • 16.30 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Designed for 120 Volt breakers that have holes in the switch tongue.
  • Quick and easy - just snap in place and apply lock.
  • Dual cavities accommodate different size switch tongues.
  • Dimensions: 8.38 x 54.36 x 30.23 mm (D x W x H)
Specifications
Specifications
Circuit Breaker Lockout
Red
Single Pole Circuit Breaker Lockout
1 Ea.