missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Nylon Small Ball Valve - Shut Off

The single-piece, easy to use design, makes this standard ball valve lockouts the ideal tool for securing your quarter-turn ball valves in the OFF position. Holes for 7 mm padlock shackles. For use on ball valve sizes from 6.35 mm to 25.40 mm

Brand:  Brady™ 065666

Product Code. 10225044

  • 29.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Holes for 7 mm padlock shackles.
  • For use on ball valve sizes from 6.35 mm to 25.40 mm.
Specifications

Specifications

Small Ball Valve Shutoff
Red
OSHA 1910.147
78.74 x 93.98 x 57.15 mm
2.25 in.
1 Ea.
93.98 mm
BALL VALVE LO - SMALL 9/32 in. NYLON
Nylon
Small Brady Ball Valve Lockout
Lockout Tagout
57.15 mm
3.7 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.