Learn More
Brady™ Nylon Thermal Transfer Printer Labels
For smooth, textured/rough or Highly textured/Low surface energy surfaces.
Brand:  Brady™ 217067
 View more versions of this product
Product Code. 11819734
  • 256.00 € / 1 roll
Estimated Shipment: 29-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Permanent acrylic adhesive
  • Matt finish
  • Nylon cloth labels
Specifications
Specifications
UL Recognized, CSA Accepted, Halogen Free (DIN VDE 0472/part 815)
Thermal Transfer Printable Labels
Yellow
Nylon
Thermal Transfer Printer Label
BBP11, PR200Plus, PR300Plus, PR600Plus, BP1244, BP1344, BP4000, BP4320
3
3000/Pk.
9.53 mm dia.
0.041 mm
9.53 mm
BPT-602-499YL
Acrylic, Permanent
Matte
Thermal Transfer
Laboratory identification
12.53 mm
3000/Pk.
Round
40 to 90°C
13.53 mm