missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ Nylon Utility Brushes

These low-cost, quality brushes are perfect for general-purpose cleaning. Cole-Parmer™ Utility Brushes feature a white plastic handle that won't support bacteria or hold water.

Brand:  Cole-Parmer™ 84550-31

19.58 EUR valid until 2025-02-13
Use promo code "24130" to get your promotional price.


 View more versions of this product

Product Code. 11755229

  • 20.80 € / Each
Estimated Shipment: 14-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Durable brushes made with nylon bristles and a hard plastic handle
Specifications

Specifications

Utility Brush
Plastic
130 mm
140 mm
1 Ea.
Nylon
White
General-purpose cleaning
500 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.