missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Nystatin, anti-fungal agent
Click to view available options
Quantity:
20 mL
Unit Size:
20mL
Description
Gibco™ Nystatin is an anti-fungal agent produced by Streptomyces noursei and Streptomyces aureus. Gibco™ Nystatin is a suspension used for the prevention of fungal contamination of cell cultures when diluted 1:100 in cell culture medium. We offer a variety of antibiotics and antimycotics for cell culture applications.
Product Use
For Research Use Only: Not intended for animal or human diagnostic or therapeutic use.
Product Use
For Research Use Only: Not intended for animal or human diagnostic or therapeutic use.
Specifications
Specifications
| Content And Storage | –5 to –20°C |
| Description | Nystatin, 20mL, –5 to –20°C Storage Temperature, Anti-Mycotic, Nystatin Agent, Anti-Mycotic Reagent Type, 18 Months Shelf Life, Gibco™ Nystatin is an Anti-Fungal Agent Produced by Streptomyces Noursei and Streptomyces Aureus |
| Form | Liquid |
| Product Type | Antifungal |
| For Use With (Application) | Mammalian cell culture, mammalian cell culture contamination |
| Quantity | 20 mL |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction