Learn More
O-(2,3,4,5,6-Pentafluorobenzyl)hydroxylamine hydrochloride, 99+%, Thermo Scientific Chemicals
Description
O-(2,3,4,5,6-Pentafluorobenzyl)hydroxylamine hydrochloride is used in the preparation of oximes of steroids having keto group. It is used as a derivatization reagent in the determination of thromboxane B2, prostaglandins, amygdalin and a variety of aldehydes, ketones and acids. It is a versatile reagent utilized for the detection of carbonyl-containing compounds by gas chromatography (GC), gas chromatography mass spectrometry (GC-MS) and other methods. As a derivatization reagent, it is also used for EC/LC analysis of keto-steroids and carbohydrates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
O-(2,3,4,5,6-Pentafluorobenzyl)hydroxylamine hydrochloride is used in the preparation of oximes of steroids having keto group. It is used as a derivatization reagent in the determination of thromboxane B2, prostaglandins, amygdalin and a variety of aldehydes, ketones and acids. It is a versatile reagent utilized for the detection of carbonyl-containing compounds by gas chromatography (GC), gas chromatography mass spectrometry (GC-MS) and other methods. As a derivatization reagent, it is also used for EC/LC analysis of keto-steroids and carbohydrates.
Solubility
Soluble in water.
Notes
Incompatible with strong oxidizing agents.
Specifications
Specifications
Melting Point | ∼215°C (sublimation) |
Quantity | 1 g |
Beilstein | 4031190 |
Solubility Information | Soluble in water. |
Formula Weight | 249.57 |
Percent Purity | ≥99% |
Chemical Name or Material | O-(2,3,4,5,6-Pentafluorobenzyl)hydroxylamine hydrochloride |
Safety and Handling
- O-(2,3,4,5,6-Pentafluorobenzyl)hydroxylamine hydrochloride
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- MIXTURE LIST-Contains : O-(2,3,4,5,6-Pentafluorobenzyl)hydroxylamine hydrochloride
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.