missing translation for 'onlineSavingsMsg'
Learn More

O-(6-Chloro-1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium tetrafluoroborate, 99+%, Thermo Scientific Chemicals

Product Code. 15425275
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 330641-16-2
Molecular Formula C11H15BClF4N5O
Molecular Weight (g/mol) 355.53
MDL Number MFCD04973270
InChI Key GBGVQFJZGHBZMC-UHFFFAOYSA-N
Synonym tctu, o-6-chlorobenzotriazol-1-yl-n,n,n',n'-tetramethyluronium tetrafluoroborate, n,n,n',n'-tetramethyl-o-6-chloro-1h-benzotriazol-1-yl uronium tetrafluoroborate, o-6-chlorobenzotriazol-1-yl-n,n,n',n'-tetramethyluronium tetrafluoroborat e, 6-chlorobenzotriazol-1-yl oxy-dimethylamino methylene-dimethyl-ammonium tetrafluoroborate, tctu; 1-bis-dimethylamino-methylene-5-chloro-3-oxy-1h-benzotriazol-1-ium tetrafluoroborate, 6-chloro-1,2,3-benzotriazol-1-yl oxy dimethylamino methylidene dimethylazanium tetrafluoroborate, 1h-benzotriazolium, 1-bis dimethylamino methylene-5-chloro-, 3-oxide tetrafluoroborate 1-1:1, acmc-1agto, ksc569q6h
PubChem CID 24898642
IUPAC Name [(6-chlorobenzotriazol-1-yl)oxy-(dimethylamino)methylidene]-dimethylazanium;tetrafluoroborate
SMILES F[B-](F)(F)F.CN(C)C(ON1N=NC2=CC=C(Cl)C=C12)=[N+](C)C

Specifications

Melting Point 198°C to 204°C
Quantity 25 g
Formula Weight 355.53
Percent Purity ≥99%
Chemical Name or Material O-(6-Chloro-1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium tetrafluoroborate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.