missing translation for 'onlineSavingsMsg'
Learn More
Learn More
O-(7-Aza-1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
O-(7-Aza-1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate is a coupling reagent and used as an additive in peptide synthesis., It is also involved efficiently to speed up the coupling process and reduces the loss of chiral integrity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
O-(7-Aza-1H-benzotriazol-1-yl)-N,N,N′,N′-tetramethyluronium hexafluorophosphate is a coupling reagent and used as an additive in peptide synthesis., It is also involved efficiently to speed up the coupling process and reduces the loss of chiral integrity.
Solubility
Soluble in acetonitrile. Insoluble in water.
Notes
Moisture sensitive. Incompatible with oxidizing agents and highly acidic or alkaline materials.
O-(7-Aza-1H-benzotriazol-1-yl)-N,N,N′,N′-tetramethyluronium hexafluorophosphate is a coupling reagent and used as an additive in peptide synthesis., It is also involved efficiently to speed up the coupling process and reduces the loss of chiral integrity.
Solubility
Soluble in acetonitrile. Insoluble in water.
Notes
Moisture sensitive. Incompatible with oxidizing agents and highly acidic or alkaline materials.
Specifications
Melting Point | ∼185°C (decomposition) |
Quantity | 1 g |
Sensitivity | Moisture sensitive |
Solubility Information | Soluble in acetonitrile. Insoluble in water. |
Formula Weight | 380.23 |
Percent Purity | 99% |
Chemical Name or Material | O-(7-Aza-1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'