missing translation for 'onlineSavingsMsg'
Learn More

o-Anisaldehyde, 98%

Product Code. 10255290 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
5g
This item is not returnable. View return policy

Product Code. 10255290

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 135-02-4 | C8H8O2 | 136.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 135-02-4
Molecular Formula C8H8O2
Molecular Weight (g/mol) 136.15
MDL Number MFCD00003308
InChI Key PKZJLOCLABXVMC-UHFFFAOYSA-N
Synonym o-anisaldehyde, benzaldehyde, 2-methoxy, 2-anisaldehyde, o-methoxybenzaldehyde, 6-methoxybenzaldehyde, salicylaldehyde methyl ether, 2-methoxybenzenecarboxaldehyde, benzaldehyde, o-methoxy, 2-methoxyphenylformaldehyde, formylanisole, o
PubChem CID 8658
IUPAC Name 2-methoxybenzaldehyde
SMILES COC1=CC=CC=C1C=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 34.0°C to 40.0°C
Color Brown to Yellow
Boiling Point 238.0°C
Flash Point 117°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3OC6H4CHO
Refractive Index 1.5608
Quantity 100 g
Beilstein 08, IV, 180
Merck Index 10, 5866
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol
Formula Weight 136.15
Percent Purity 98%
Physical Form Crystalline Low Melting Solid
Chemical Name or Material o-Anisaldehyde, 98%
Show More Show Less
compliance-icons
Product Identifier
  • o-Anisaldehyde
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.