missing translation for 'onlineSavingsMsg'
Learn More
O-Benzylphospho-N-Fmoc-L-serine, 95%, Thermo Scientific Chemicals Product Code.: 15428707

O-Benzylphospho-N-Fmoc-L-serine, 95%, Thermo Scientific Chemicals

Product Code. 15428707
1 g
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy

Product Code. 15428707

Brand: Thermo Scientific Alfa Aesar H52168.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

It can be applied in the synthesis of phosphopeptides. This derivative can be introduced using standard activation methods, such as PyBOP and TBTU. The monoprotected phosphoserine residue once incorporated is stable to piperidine. Using this reagent, even peptides containing multiple phosphorylation sites can be prepared efficiently by standard Fmoc SPPS methods. β-piperidinylalanine formation has been shown to occur during Fmoc deprotection of N-terminal Ser(PO(OBzl)OH), particularly under microwave conditions. This side reaction can be eliminated by using cyclohexylamine or DBU just for this Fmoc deprotection step .

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It can be applied in the synthesis of phosphopeptides. This derivative can be introduced using standard activation methods, such as PyBOP and TBTU. The monoprotected phosphoserine residue once incorporated is stable to piperidine. Using this reagent, even peptides containing multiple phosphorylation sites can be prepared efficiently by standard Fmoc SPPS methods. β-piperidinylalanine formation has been shown to occur during Fmoc deprotection of N-terminal Ser(PO(OBzl)OH), particularly under microwave conditions. This side reaction can be eliminated by using cyclohexylamine or DBU just for this Fmoc deprotection step .

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 158171-14-3
Molecular Formula C25H24NO8P
Molecular Weight (g/mol) 497.44
MDL Number MFCD00797869
InChI Key ZBPUWGDUVAAWJY-QHCPKHFHSA-N
Synonym fmoc-o-benzylphospho-l-serine, fmoc-ser hpo3bzl-oh, fmoc-o-benzyl-l-phosphoserine, fmoc-ser po3bzlh-oh, 2s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-benzyloxy hydroxy phosphoryl oxy propanoic acid, 2s-3-benzyloxy hydroxy phosphoryl oxy-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid, pubchem20516, fmoc-l-ser po obzl oh-oh, n-fmoc-o-benzylphospho-l-serine, fmoc-ser po3bzlh-oh hplcShow MoreShow Less
PubChem CID 11005563
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-[hydroxy(phenylmethoxy)phosphoryl]oxypropanoic acid
SMILES C1=CC=C(C=C1)COP(=O)(O)OCC(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24

Specifications

Quantity 1 g
Beilstein 7060483
Solubility Information Slightly soluble in water.
Formula Weight 497.44
Percent Purity 95%
Chemical Name or Material O-Benzylphospho-N-Fmoc-L-serine

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberX26I028 Certificate TypeCertificate of Analysis Date23/01/2023 Catalog NumberH52168.MD,H52168.06,H52168.03
Lot NumberM25D013 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH52168.MD,H52168.06,H52168.03
Lot NumberN25A018 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH52168.MD,H52168.06,H52168.03
Lot NumberY01C052 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH52168.MD,H52168.06,H52168.03
Lot NumberX04H024 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH52168.MD,H52168.06,H52168.03

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
O-Benzylphospho-N-Fmoc-L-serine, 95%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.