Learn More
o-Cresol, 99%, AcroSeal™, Thermo Scientific Chemicals
20.20 € - 49.55 €
Chemical Identifiers
CAS | 95-48-7, 108-95-2 |
---|---|
Molecular Formula | C7H8O |
Molecular Weight (g/mol) | 108.14 |
MDL Number | MFCD00002226 |
InChI Key | QWVGKYWNOKOFNN-UHFFFAOYSA-N |
Synonym | o-cresol, 2-hydroxytoluene, o-methylphenol, 2-cresol, phenol, 2-methyl, orthocresol, o-cresylic acid, o-oxytoluene, o-toluol, o-hydroxytoluene |
PubChem CID | 335 |
ChEBI | CHEBI:28054 |
IUPAC Name | 2-methylphenol |
SMILES | CC1=CC=CC=C1O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10503648
|
Thermo Scientific Acros
405731000 |
100 mL | AcroSeal™ Glass Bottle |
20.20 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10278127
|
Thermo Scientific Acros
405738000 |
800 mL | AcroSeal™ Glass Bottle |
49.55 €
800mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
95-48-7, 108-95-2 | |
108.14 | |
QWVGKYWNOKOFNN-UHFFFAOYSA-N | |
335 | |
2-methylphenol |
C7H8O | |
MFCD00002226 | |
o-cresol, 2-hydroxytoluene, o-methylphenol, 2-cresol, phenol, 2-methyl, orthocresol, o-cresylic acid, o-oxytoluene, o-toluol, o-hydroxytoluene | |
CHEBI:28054 | |
CC1=CC=CC=C1O |
Specifications
95-48-7, 108-95-2 | |
100.0 | |
Amber to Colorless | |
191.0°C | |
Authentic | |
C7H8O | |
MFCD00002226 | |
15,2564 | |
o-cresol, 2-hydroxytoluene, o-methylphenol, 2-cresol, phenol, 2-methyl, orthocresol, o-cresylic acid, o-oxytoluene, o-toluol, o-hydroxytoluene | |
QWVGKYWNOKOFNN-UHFFFAOYSA-N | |
2-methylphenol | |
335 | |
4.75 mPa.s (35°C) | |
99% | |
o-Cresol |
98.5 | |
30.0°C to 32.0°C | |
1.0400g/mL | |
81°C | |
98.5% min. (GC) | |
CH3C6H4OH | |
06,349 | |
1.04 | |
Solubility in water: 20g/L (20°C). Other solubilities: soluble in alcohol,ether and chloroform | |
CC1=CC=CC=C1O | |
108.14 | |
CHEBI:28054 | |
108.14 | |
Liquid or Low Melting Solid |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Harmful to aquatic life with long lasting effects.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN:
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 202-423-8
Recommended Storage : Darkens on storage and exposure to light and air
- o-Cresol
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
- MIXTURE LIST-Contains : o-Cresol
RUO – Research Use Only