missing translation for 'onlineSavingsMsg'
Learn More
Learn More
o-Cresolphthalein Complex, Honeywell Fluka™
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass Bottle
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
Melting Point | 179°C to 182°C |
Boiling Point | >260°C |
UN Number | NONH for all modes of transport |
Beilstein | 381994 |
Formula Weight | 636.6g/mol |
Odor | Unpleasant |
Packaging | Glass Bottle |
Color | Yellow |
Quantity | 5 g |
Physical Form | Powder |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction