missing translation for 'onlineSavingsMsg'
Learn More

o-Dianisidine dihydrochloride, 98%, Thermo Scientific Chemicals

Product Code. 11413397
Click to view available options
Quantity:
1 g
5 g

It is suitable for use in ELISA procedures. In neurohistochemistry o-Dianisidine dihydrochloride has demonstrated perikaryal labeling as well as retrograde and anterograde transport to the primary olfactory cortex. It has also been used for a paper based-micro fluidic device to semi-quantitatively measure lactate, uric acid, and glucose. 3,3'-Dimethoxybenzidine is used principally as an intermediate in the production of commercial bisazobiphenyl dyes for coloring textiles, paper, plastic, rubber, and leather.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is suitable for use in ELISA procedures. In neurohistochemistry o-Dianisidine dihydrochloride has demonstrated perikaryal labeling as well as retrograde and anterograde transport to the primary olfactory cortex. It has also been used for a paper based-micro fluidic device to semi-quantitatively measure lactate, uric acid, and glucose. 3,3′-Dimethoxybenzidine is used principally as an intermediate in the production of commercial bisazobiphenyl dyes for coloring textiles, paper, plastic, rubber, and leather.

Solubility
Soluble in water (50 mg/ml). It dissolves in ethyl alcohol ,acid and in organic solvent.

Notes
Store away from oxidizing agents. Keep the container tightly sealed and store in cool, dry condition in well ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20325-40-0
Molecular Formula C14H18Cl2N2O2
Molecular Weight (g/mol) 317.21
MDL Number MFCD00012488
InChI Key UXTIAFYTYOEQHV-UHFFFAOYSA-N
Synonym 3,3'-dimethoxybenzidine dihydrochloride, o-dianisidine dihydrochloride, 3,3'-dimethoxy-1,1'-biphenyl-4,4'-diamine dihydrochloride, unii-qv96qa6ukk, dianisidine dihydrochloride, ccris 998, o-dianisidine hcl, qv96qa6ukk, c.i. disperse black 6 dihydrochloride
PubChem CID 62311
IUPAC Name 4-(4-amino-3-methoxyphenyl)-2-methoxyaniline;dihydrochloride
SMILES COC1=C(C=CC(=C1)C2=CC(=C(C=C2)N)OC)N.Cl.Cl

Specifications

Melting Point 268°C (decomposition)
Assay Percent Range 99%
Quantity 5 g
Beilstein 3917996
Solubility Information Soluble in water (50mg/ml). It dissolves in ethyl alcohol ,acid and in organic solvent.
Formula Weight 317.21
Percent Purity 99%
Chemical Name or Material o-Dianisidine dihydrochloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'