missing translation for 'onlineSavingsMsg'
Learn More

O-(endo-5-Norbornene-2,3-dicarboximido)-N,N,N',N'-tetramethyluronium tetrafluoroborate, 98+%, Thermo Scientific Chemicals

Product Code. 15425265
25 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 125700-73-4
Molecular Formula C14H20BF4N3O3
Molecular Weight (g/mol) 365.14
MDL Number MFCD00167753
InChI Key DMTWWFCCBWWXSA-UHFFFAOYNA-N
Synonym 2-endo-5-norbornene-2,3-dicarboxymido-1,1,3,3-tetramethyluronium tetrafluoroborate, dimethylamino 1r,2s,6r,7s-3,5-dioxo-4-azatricyclo 5.2.1.0_,? dec-8-en-4-yl oxy methylidene dimethylazanium tetrafluoroborate
PubChem CID 91634054
IUPAC Name [(dimethylamino)({3,5-dioxo-4-azatricyclo[5.2.1.0²,⁶]dec-8-en-4-yl}oxy)methylidene]dimethylazanium; tetrafluoroboranuide
SMILES F[B-](F)(F)F.CN(C)C(ON1C(=O)C2C3CC(C=C3)C2C1=O)=[N+](C)C

Specifications

Melting Point 216°C to 219°C
Quantity 25 g
Sensitivity Light and moisture sensitive
Formula Weight 365.13
Percent Purity ≥98%
Chemical Name or Material O-(endo-5-Norbornene-2,3-dicarboximido)-N,N,N',N'-tetramethyluronium tetrafluoroborate
compliance-icons
Product Identifier
  • O-(endo-5-Norbornene-2,3-dicarboximido)-N,N,N',N'-tetramethyluronium tetrafluoroborate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : O-(endo-5-Norbornene-2,3-dicarboximido)-N,N,N',N'-tetramethyluronium tetrafluoroborate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
O-(endo-5-Norbornene-2,3-dicarboximido)-N,N,N',N'-tetramethyluronium tetrafluoroborate, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.