missing translation for 'onlineSavingsMsg'
Learn More

o-Fluorotoluene, 99%

Product Code. 10605352 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Unit Size:
100g
10g
2.5kg
500g
This item is not returnable. View return policy

Product Code. 10605352

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 95-52-3 | C7H7F | 110.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-52-3
MDL Number MFCD00000322
InChI Key MMZYCBHLNZVROM-UHFFFAOYSA-N
Synonym 2-fluorotoluene, o-fluorotoluene, benzene, 1-fluoro-2-methyl, toluene, o-fluoro, 1-methyl-2-fluorobenzene, unii-l55xkb3k7h, 1-fluoro-2-methyl-benzene, ortho-fluorotoluene, fluorotoluene, l55xkb3k7h
PubChem CID 7241
IUPAC Name 1-fluoro-2-methylbenzene
SMILES CC1=CC=CC=C1F

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -62.0°C
Color Colorless to Yellow
Density 1.0000g/mL
Boiling Point 113.0°C to 114.0°C
Flash Point 8°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Molecular Formula C7H7F
Linear Formula CH3C6H4F
Refractive Index 1.4720 to 1.4750
Quantity 100 g
Beilstein 05, 290
Merck Index 15, 4212
Specific Gravity 1
Solubility Information Solubility in water: insoluble. Other solubilities: miscible with alcohol and ether
Molecular Weight (g/mol) 110.13
Formula Weight 110.13
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material o-Fluorotoluene, 99+%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • o-Fluorotoluene
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H335-May cause respiratory irritation.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.