missing translation for 'onlineSavingsMsg'
Learn More

o-Phenetidine, 99%, Thermo Scientific Chemicals

Product Code. 10246290
Click to view available options
Quantity:
5 mL
500 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 94-70-2
Molecular Formula C8H11NO
Molecular Weight (g/mol) 137.18
MDL Number MFCD00007689
InChI Key ULHFFAFDSSHFDA-UHFFFAOYSA-N
Synonym o-phenetidine, 2-aminophenetole, benzenamine, 2-ethoxy, o-ethoxyaniline, 2-ethoxybenzenamine, o-aminophenetole, 2-ethoxyphenylamine, ethoxyaniline, 2-phenetidine, benzenamine, ar-ethoxy
PubChem CID 7203
IUPAC Name 2-ethoxyaniline
SMILES CCOC1=CC=CC=C1N

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -20.0°C
Color Red
Density 1.0500g/mL
Boiling Point 230.0°C to 233.0°C
Flash Point 80°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula C2H5OC6H4NH2
Refractive Index 1.5540 to 1.5560
Quantity 5 mL
Merck Index 15, 7341
Specific Gravity 1.05
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol and ether
Formula Weight 137.18
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material o-Phenetidine, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • o-Phenetidine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H373-May cause damage to organs.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.