missing translation for 'onlineSavingsMsg'
Learn More
Learn More
O-Phthalaldehyde, Extra Pure, Fisher Chemical
Click to view available options
Quantity:
5 g
Packaging:
Glass Jar
Specifications
Specifications
CAS | 643-79-8 |
Melting Point | 54°C |
Color | Yellow |
pH | 7 |
Boiling Point | 83°C |
Packaging | Glass Jar |
Molecular Formula | C8H6O2 |
MDL Number | MFCD00003335 |
Quantity | 5 g |
Synonym | o-phthalaldehyde, o-phthaldialdehyde, benzene-1,2-dicarboxaldehyde, 1,2-benzenedicarboxaldehyde, phthaldialdehyde, phthalic aldehyde, phthalic dialdehyde, phthalyldicarboxaldehyde, ortho-phthalaldehyde, o-phthaldehyde |
Show More |
Safety and Handling
Product Identifier
- o-Phthalaldehyde
- Danger
- Acute toxicity Category 3
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- Skin sensitiser Category 1
- ACUTE AQUATIC Acute 1
- H314-Causes severe skin burns and eye damage.
- H317-May cause an allergic skin reaction.
- H301-Toxic if swallowed.
- H400-Very toxic to aquatic life.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P273-Avoid release to the environment.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'