missing translation for 'onlineSavingsMsg'
Learn More

o-Tolidine, 95%, pract, Thermo Scientific Chemicals

Product Code. 10687031
Click to view available options
Quantity:
25 g
100 g
250 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 119-93-7
MDL Number MFCD00014773
InChI Key NUIURNJTPRWVAP-UHFFFAOYSA-N
Synonym o-tolidine, 3,3'-dimethylbenzidine, orthotolidine, diaminoditolyl, 2-tolidine, diaminotolyl, bianisidine, 3,3'-tolidine, 3,3'-dimethylbiphenyl-4,4'-diamine, 4,4'-bi-o-toluidine
PubChem CID 8413
ChEBI CHEBI:34320
IUPAC Name 4-(4-amino-3-methylphenyl)-2-methylaniline
SMILES CC1=C(C=CC(=C1)C2=CC(=C(C=C2)N)C)N

Specifications

Melting Point 125.0°C to 132.0°C
Color Beige or Brown to Gray
Boiling Point 300.5°C
Flash Point 244°C
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Molecular Formula C14H16N2
Quantity 25 g
Beilstein 13, IV, 419
Merck Index 15, 9674
Molecular Weight (g/mol) 212.29
Formula Weight 212.29
Percent Purity 95%
Physical Form Powder
Chemical Name or Material o-Tolidine, (pract), 95%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • o-Tolidine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Carcinogenicity Category 1B
Hazard Statement
  • H350-May cause cancer.
  • H411-Toxic to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P273-Avoid release to the environment.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.