missing translation for 'onlineSavingsMsg'
Learn More

o-Tolidine, 98%, Thermo Scientific Chemicals

Product Code. 11473428
Click to view available options
Quantity:
25 g
100 g

O-Tolidine is widely used in dye production. It is particularly used in textile, leather and paper industries. It can be used as an indicator in analytical, clinical and forensic chemistry. It is utilized for the determination of gold and chlorine level in swimming pool water.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
O-Tolidine is widely used in dye production. It is particularly used in textile, leather and paper industries. It can be used as an indicator in analytical, clinical and forensic chemistry. It is utilized for the determination of gold and chlorine level in swimming pool water.

Solubility
Slightly soluble in water.

Notes
Light and air sensitive. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 119-93-7
Molecular Formula C14H16N2
Molecular Weight (g/mol) 212.296
MDL Number MFCD00014773
InChI Key NUIURNJTPRWVAP-UHFFFAOYSA-N
Synonym o-tolidine, 3,3'-dimethylbenzidine, orthotolidine, diaminoditolyl, 2-tolidine, diaminotolyl, bianisidine, 3,3'-tolidine, 3,3'-dimethylbiphenyl-4,4'-diamine, 4,4'-bi-o-toluidine
PubChem CID 8413
ChEBI CHEBI:34320
IUPAC Name 4-(4-amino-3-methylphenyl)-2-methylaniline
SMILES CC1=C(C=CC(=C1)C2=CC(=C(C=C2)N)C)N

Specifications

Melting Point 128°C to 132°C
Boiling Point 300°C to 301°C
Flash Point 244°C (471°F)
Quantity 25 g
UN Number UN3077
Sensitivity Light sensitive
Merck Index 14,9514
Solubility Information Slightly soluble in water.
Formula Weight 212.3
Percent Purity 98%
Chemical Name or Material o-Tolidine
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • o-Tolidine
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 1A
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
  • H350-May cause cancer.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P273-Avoid release to the environment.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
  • P501-Dispose of contents/container to in accordance with local/regional/national/international regulations..

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.