missing translation for 'onlineSavingsMsg'
Learn More

o-Toluoyl chloride, 98+%, Thermo Scientific Chemicals

51.96 € - 197.99 €

Chemical Identifiers

CAS 933-88-0
Molecular Formula C8H7ClO
Molecular Weight (g/mol) 154.59
MDL Number MFCD00000668
InChI Key GPZXFICWCMCQPF-UHFFFAOYSA-N
Synonym o-toluoyl chloride, benzoyl chloride, 2-methyl, o-toluoylchloride, methylbenzoyl chloride, o-methylbenzoyl chloride, 2-toluoyl chloride, o-toluic acid chloride, unii-j9ykn18e7r, o-toluyl chloride, 2-methylbenzoic acid chloride
PubChem CID 70276
IUPAC Name 2-methylbenzoyl chloride
SMILES CC1=CC=CC=C1C(Cl)=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11429682
View Documents
Thermo Scientific Alfa Aesar
A10775.22
100 g
51.96 €
100g
Please to purchase this item. Need a web account? Register with us today!
11439682
View Documents
Thermo Scientific Alfa Aesar
A10775.36
500 g
197.99 €
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

o-Toluoyl chloride is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
o-Toluoyl chloride is used as a pharmaceutical intermediate.

Solubility
Reacts with water.

Notes
Keep container tightly sealed. Store under dry inert gas. Store in cool, dry conditions in well sealed containers. Incompatible with bases, active metals, alcohols,amines and oxidizing agents. It is sensitive to moisture.
Specifications

Chemical Identifiers

933-88-0
154.59
GPZXFICWCMCQPF-UHFFFAOYSA-N
70276
CC1=CC=CC=C1C(Cl)=O
C8H7ClO
MFCD00000668
o-toluoyl chloride, benzoyl chloride, 2-methyl, o-toluoylchloride, methylbenzoyl chloride, o-methylbenzoyl chloride, 2-toluoyl chloride, o-toluic acid chloride, unii-j9ykn18e7r, o-toluyl chloride, 2-methylbenzoic acid chloride
2-methylbenzoyl chloride

Specifications

933-88-0
212°C to 214°C
Pungent
1.5549
UN3265
Moisture sensitive
Reacts with water.
CC1=CC=CC=C1C(Cl)=O
154.59
154.6
o-Toluoyl chloride
1.187
76°C (168°F)
C8H7ClO
MFCD00000668
507933
o-toluoyl chloride, benzoyl chloride, 2-methyl, o-toluoylchloride, methylbenzoyl chloride, o-methylbenzoyl chloride, 2-toluoyl chloride, o-toluic acid chloride, unii-j9ykn18e7r, o-toluyl chloride, 2-methylbenzoic acid chloride
GPZXFICWCMCQPF-UHFFFAOYSA-N
2-methylbenzoyl chloride
70276
≥98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.

P210-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H227-H314

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

missing translation for 'einecsNumber' : 213-273-8

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only