missing translation for 'onlineSavingsMsg'
Learn More

o-Vanillin, 99%

Product Code. 10143680 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Unit Size:
100g
10g
2.5kg
500g
This item is not returnable. View return policy

Product Code. 10143680

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 148-53-8 | C8H8O3 | 152.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 148-53-8
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.15
MDL Number MFCD00003322
InChI Key JJVNINGBHGBWJH-UHFFFAOYSA-N
Synonym 3-methoxysalicylaldehyde, o-vanillin, 6-formylguaiacol, 2-vanillin, o-vanilline, benzaldehyde, 2-hydroxy-3-methoxy, orthovanilline, 2-hydroxy-m-anisaldehyde, 6-formyl-2-methoxyphenol, 3-methoxy-2-hydroxybenzaldehyde
PubChem CID 8991
ChEBI CHEBI:78339
IUPAC Name 2-hydroxy-3-methoxybenzaldehyde
SMILES COC1=CC=CC(C=O)=C1O

Specifications

Melting Point 40.0°C to 42.0°C
Color Brown to Yellow
Boiling Point 265.0°C to 266.0°C
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 100 g
Beilstein 08, IV, 1747
Solubility Information Solubility in water: slightly soluble. Other solubilities: very soluble in alcohol,ether,benzene,carbon,disulfide
Formula Weight 152.15
Percent Purity 99%
Physical Form Low Melting Solid
Chemical Name or Material o-Vanillin, 99%
Show More Show Less
compliance-icons
Product Identifier
  • o-Vanillin
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.