missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ Oakton™ Buffer Solution

Ideal for laboratory and field analysis

Brand:  Cole-Parmer™ 05942-44

Product Code. 11715089

  • 58.30 € / Each
Estimated Shipment: 26-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Oakton Buffer Solution, pH 7.00; 1 gallon
  • All Oakton pH buffers are traceable to NIST standard reference materials
  • Buffer solution lets you calibrate any pH instrument to ensure accuracy
  • Makes calibration easy
  • Simply adjust your instrument to the pH value of the solution
  • If unopened, shelf life is 2 years
  • Oakton pH Buffer Solutions have pH/temperature tables printed on the labels for accurate calibrations
  • Manufactured under ISO 9000 quality guidelines and traceable to NIST standards
  • If unopened, shelf life is 2 years; once opened, use pH 4 and pH 7 within 90 days, pH 10 within 30 days
  • Buffer Options:
  • pH 1.68 buffer available in clear
  • pH 4.01 buffer available in red
  • pH 6.86 buffer available in clear
  • pH 7.00 buffer available in yellow
  • pH 9.18 buffer available in clear
  • pH 10.01 buffer available in blue
  • pH 12.46 buffer available in clear
  • Buffer is colored for quick-glance recognition and does not effect accuracy or stability
As of April 2018, Oakton will be using yellow for the color-reference of the pH 7 buffer solution instead of green. Standardized against NIST-traceable pH references for meters with 0.01 resolution. Sold in individual jugs; 1-gallon bottles are also available as a case of four. Store buffers away from direct sunlight.
Specifications

Specifications

Yellow
Oakton Buffer Solution, pH 7
1 Gallon (4,000mL)
pH 7 Buffer Standard
Laboratory and field analysis
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.