Learn More
Conductivity Calibration Pouches, Oakton™
Calibrate on the go with these single-use 20mL calibration pouches. Oakton™ Conductivity Calibration Pouches are sealed to ensure freshness.
Brand:  Cole Parmer 35653-11
 View more versions of this product
Product Code. 12393780
  • 52.65 € / Pack of 20
Explore more special offers
Description
Description
Calibrate on the go with these single-use 20mL calibration pouches. Oakton™ Conductivity Calibration Pouches are sealed to ensure freshness.
  • Convenient calibration pouches are portable
  • A μS-to-ppm conversion table is printed on each pack for quick reference
  • Accuracy is ±1% at 25±C
  • Calibration pouches include a NIST-traceable calibration report supplied by the manufacture
Specifications
Specifications
Cole-Parmer Conductivity Standards
Calibration Solution
ppm NaCl: 702, ppm 442: 1000, ppm KCl: 745
20 Single-use Pouches
Conductivity Standard