Learn More
Digi-Sense™ Infrared Food Safety Thermometers
Ideal for food inspections and HACCP programs.
111.00 € - 185.00 €
Products 3
Product Code Brand Temperature Range (Metric) Price Quantity & Availability  
Product Code Brand Temperature Range (Metric) Price Quantity & Availability  
11781485
View Documents
Digi-Sense™
35625-45
-30°C to +200°C
124.00 €
Each
Estimated Shipment: 03-06-2025
to see stock.
Add to basket
11745009
View Documents
Digi-Sense™
35625-40
-18°C to +302°C N/A N/A N/A
11756088
View Documents
Digi-Sense™
3562515
-35°C to +275°C (Infrared); 0°C to 200°C (Contact Probe)
111.00 €
Each
Estimated Shipment: 17-06-2025
to see stock.
Add to basket
Description
Description
  • Can also be used in the home, office, field or lab, anywhere you need to quickly and accurately measure surface temperatures
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Digi-Sense™ Infrared Food Safety Thermometers