Learn More
pH Calibration Buffer Bottles, Oakton™
Product Code. 11750079
Click to view available options
pH:
1.68
4.01
7.00
10.00
12.46
Use these PH buffers for meters with 0.01 resolution. Oakton™ pH Calibration Buffer Bottles are standardized against NIST-traceable pH references within 0.01 pH at 77°F (25°C).
  • pH buffer solutions allow the user to adjust their system to read accurate measurements
  • Store buffers away from direct sunlight.
Order Info
Shipping weight: 1-1/4 lbs. (0.6kg).
Certifications
NIST traceable, Certified to NIST traceable standards.
Specifications
pH 10.00
Quantity 500 mL