missing translation for 'onlineSavingsMsg'
Learn More
Learn More
pH Calibration Buffer Bottles, Oakton™
Click to view available options
pH:
1.68
4.01
7.00
10.00
12.46
Description
- pH buffer solutions allow the user to adjust their system to read accurate measurements
- Store buffers away from direct sunlight.
Order Info
Shipping weight: 1-1/4 lbs. (0.6kg).
Certifications
NIST traceable, Certified to NIST traceable standards.
Specifications
Specifications
pH | 4.01 |
Quantity | 500 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction