missing translation for 'onlineSavingsMsg'
Learn More

Oakton™ Waterproof Big Display pHTestr™ 30

Versatile, compact design works in a field or laboratory setting.

Brand:  Oakton™ 35634-30

 View more versions of this product

Product Code. 13444606

  • 146.00 € / Each
Estimated Shipment: 11-02-2025
to see stock.
Explore more special offers
Add to basket

Includes: Meter Only package includes meter, electrode, four 1.5V batteries, instructions. Meter Kit package includes meter; electrode; two each of pH 4.01, 7.00, 10.00 buffers and rinse solution in single-use pouches, sample jar, batteries, instructions and hard Plastic carrying case
This item is not returnable. View return policy

Description

Description

  • Simultaneous pH and temperature display
  • Temperature measurement from 0.0° to 50.0°C
  • Suitable for most ASTM and EPA test procedures
  • Microprocessor-based, with accurate readings to 0.01pH
  • Replaceable double-junction electrode (S65290 or NC9830226) has extra reference gel for longer electrode life
  • Automatic temperature compensation
  • Push-button three-point calibration
  • Auto buffer recognition: selectable buffer sets
  • Hold function; auto-off; error messages
  • Waterproof, dustproof and floatable; certifiable to IP67 standards

Order Info

  • Ordering Information
  • Meter dimensions: 6-1/2 in.H x 1-1/2 in. dia. (16.5 x 3.8cm). Shipping weight (meter only): 4.5 oz. (125g).

Services

One-year warranty.

Specifications

Specifications

Meter, electrode, four 1.5V batteries, instructions
±0.01
-1 to 15
1 to 3
LCD
IP-67
±0.1 pH
-1 to 15 pH
0°C to 50°C
1.5 in.
±1°C
Hold function, auto-off, error messages, waterproof, dustproof and floatable
0.01
0.1°C
USA, NIST
1 Year
0.1pH
32.0°F to 122.0°F
16.5 cm
3.8 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.