missing translation for 'onlineSavingsMsg'
Learn More

SCHOTT AG Lighting and Imaging™ Objective Adapter for Fiber Optic or LED Ring Lights

The Objective Adapter for Fiber Optic or LED Ring Lights allows precise positioning of Ring Light.

Brand:  SCHOTT AG Lighting and Imaging™ 157569

 View more versions of this product

Product Code. 12876543

  • 160.00 € / Each
Estimated Shipment: 13-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Objective adapter for fiber optic or LED Ring Lights.
  • Adapters for a variety of objective series.
  • For objectives with 50 mm, 58 mm or 60 mm diameter.
  • Extension adapter for darkfields.
  • Available for working distances 30 to 50 mm or 50 to 80 mm or 80 to 110 mm.
  • Precise positioning of Ring Light.
Specifications

Specifications

Adapter
Extension adapter for darkfield ringlights, working distance 30 to 50 mm
Stereo Microscopy Illumination
Fiber optic or LED Darkfield Ringlight
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.