missing translation for 'onlineSavingsMsg'
Learn More

Notrax Oct-O-Flex Beveled™ Stylish Outdoor Rubber Entrance Mat

The Oct-O-Flex Beveled™ outdoor rubber mat is known for its octagonal ring design with small drainage holes and smooth surface making this next generation mat EU compliant for public entrances and the ideal mat for wheeled access.

Brand:  Notrax 599B7090OB

 View more versions of this product

Product Code. 17762238

  • 56.00 € / Each
Estimated Shipment: 26-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • The Oct-O-Flex Beveled™ outdoor rubber mat is known for its octagonal ring design with small drainage holes and smooth surface making this next generation mat EU compliant for public entrances and the ideal mat for wheeled access (carts, wheelchairs, suitcases, etc)
  • Moulded edges on all four sides are bevelled to allow easy access onto the mat and prevent tripping
  • Made from a durable rubber compound that withstands the heaviest of foot traffic, Oct-O-Flex™ mats are easy to clean and practically weather proof
  • The Oct-O-Flex Beveled™ has excellent adherence and adapts to any surface while the raised studs underneath allow water to easily drain through, keeping the surface clean and dry
Specifications

Specifications

Black
90 cm
Industrial and Commercial Matting
35 in.
Natural Rubber
70 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.