missing translation for 'onlineSavingsMsg'
Learn More

Notrax Oct-O-Mat™ Classic Honeycomb Entrance Mat

The traditional octagonal Oct-O-Mat™ outdoor rubber mat is known for its large holes that provide traction and excellent drainage in extreme weather conditions, and scrape shoes to stop dirt, grit and grime from entering the building.

Brand:  Notrax 564S3248OB

 View more versions of this product

Product Code. 17702228

  • 73.00 € / Each
Estimated Shipment: 09-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • The traditional octagonal Oct-O-Mat™ outdoor rubber mat is known for its large holes that provide traction and excellent drainage in extreme weather conditions and scrape shoes to stop dirt, grit and grime from entering the building
  • Made from a durable rubber compound, these heavy duty mats withstand intense foot traffic, are easy to clean and practically weather proof
  • The resilient rubber offers excellent adherence and adapts to any surface while the raised knobs underneath allow water to easily flow away in all directions, keeping the surface clear
Specifications

Specifications

Black
120 cm
Industrial and Commercial Matting
47 in.
Natural Rubber
80 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers