missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Octacarbonyldicobalt, stab. with 1-5% hexane, Thermo Scientific Chemicals
Click to view available options
Quantity:
2.5 g
25 g
100 g
Description
As a catalyst in oxo process, and in high-purity cobalt saltsOctacarbonyldicobalt is used as a catalyst for hydroformylation. It plays an important role in promoting both the Nicholas reaction and the Pauson-Khand reaction. It acts as a reagent and catalyst in organometallic chemistry and organic synthesis. It is involved as an alkyne protecting group.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | ∼51°C (decomposition) |
Color | Red to Orange |
Density | 1.73 g/mL |
Flash Point | -13°C (-9°F) |
Linear Formula | Co2(CO)8 |
Quantity | 2.5 g |
UN Number | UN3124 |
Sensitivity | Air sensitive |
Solubility Information | Insoluble in water; Soluble in alcohol,ether and carbon disulfideSoluble in ether,naphtha and carbon disulfide; Slightly soluble in alcohol; Insoluble in water. |
Formula Weight | 341.95 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'