Learn More
Macherey-Nagel™ Octadecyl Phase Polygosil™
Brand:  Macherey-Nagel™ 711570.10
 View more versions of this product
Product Code. 10318441
  • 210.00 € / 10g
Estimated Shipment: 16-12-2024
to see stock.
Explore more special offers
Add to basket
Specifications
7 μm
Liquid Chromatography
Polygosil C18 Phases
100 Å
10 g