Learn More
Macherey-Nagel™ Octadecyl Phase Polygosil™ C18
Brand:  Macherey-Nagel™ 711340.100
 View more versions of this product
Product Code. 10194591
  • 1730.00 € / 100g
Estimated Shipment: 13-02-2025
to see stock.
Explore more special offers
Add to basket
Specifications
7 μm
Liquid Chromatography
Polygosil C18 Phases
60 Å
100 g