Learn More
Macherey-Nagel™ Octadecyl Phase Polygosil™ C18
Brand:  Macherey-Nagel™ 711992.10
 View more versions of this product
Product Code. 10296401
  • 235.00 € / 10g
Estimated Shipment: 27-02-2025
to see stock.
Explore more special offers
Add to basket
Specifications
7 μm
Liquid Chromatography
Polygosil C18 Phases
1000 Å
10 g