Learn More
Macherey-Nagel™ Octadecyl Phase Polygosil™ C18
Brand:  Macherey-Nagel™ 711350.10
 View more versions of this product
Product Code. 10419351
  • 188.00 € / 10g
Estimated Shipment: 30-01-2025
to see stock.
Explore more special offers
Add to basket
Specifications
10 μm
Liquid Chromatography
Polygosil C18 Phases
60 Å
10 g