Learn More
Macherey-Nagel™ Octadecyl Phase Polygosil™ C18
Brand:  Macherey-Nagel™ 711992.100
 View more versions of this product
Product Code. 10515781
  • 2045.00 € / 100g
Estimated Shipment: 30-01-2025
to see stock.
Explore more special offers
Add to basket
Specifications
7 μm
Liquid Chromatography
Polygosil C18 Phases
1000 Å
100 g