missing translation for 'onlineSavingsMsg'
Learn More
Octadecylamine, 90%, tech., Thermo Scientific Chemicals Product Code.: 10785231

Octadecylamine, 90%, tech., Thermo Scientific Chemicals

Product Code. 10785231
500 g, Glass bottle
Click to view available options
Quantity:
500 g
1 kg
5 kg
Packaging:
Glass bottle
Plastic drum
This item is not returnable. View return policy

Product Code. 10785231

Brand: Thermo Scientific Acros 129325000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 124-30-1
Molecular Formula C18H39N
Molecular Weight (g/mol) 269.52
MDL Number MFCD00008159
InChI Key REYJJPSVUYRZGE-UHFFFAOYSA-N
Synonym octadecylamine, 1-octadecanamine, stearylamine, 1-octadecylamine, octadecanamine, stearamine, 1-aminooctadecane, n-octadecylamine, n-stearylamine, armofilm
PubChem CID 15793
ChEBI CHEBI:63866
IUPAC Name octadecan-1-amine
SMILES CCCCCCCCCCCCCCCCCCN

Specifications

CAS Min % 88.0
CAS Max % 100.0
Melting Point 50.0°C to 60.0°C
Color White
Boiling Point 232.0°C (32.0 mmHg)
Flash Point 148°C
Infrared Spectrum Authentic
Assay Percent Range 88% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)17NH2
Quantity 500 g
Beilstein 04, IV, 825
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in hydrocarbons
Viscosity 4.17 mPa.s (20°C)
Formula Weight 269.51
Percent Purity 90%
Grade Technical
Concentration or Composition (by Analyte or Components) Primary amine (MW 269):> 98%, Sec. + tert. amine (MW 521): < 2%
Physical Form Prills
Chemical Name or Material Octadecylamine
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0466227 Certificate TypeCertificate of Analysis Date01/10/2024 Catalog Number129320010,129325000,129320050
Lot NumberA0463960 Certificate TypeCertificate of Analysis Date28/06/2024 Catalog Number129320010,129325000,129320050
Lot NumberA0423279 Certificate TypeCertificate of Analysis Date16/05/2023 Catalog Number129320010,129325000,129320050
Lot NumberA0446581 Certificate TypeCertificate of Analysis Date24/10/2022 Catalog Number129320010,129325000,129320050
Lot NumberA0442195 Certificate TypeCertificate of Analysis Date17/05/2022 Catalog Number129320010,129325000,129320050
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Octadecylamine
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • ACUTE AQUATIC Acute 1
  • Aspiration toxicity Category 1
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H315-Causes skin irritation.
  • H373-May cause damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
  • H304-May be fatal if swallowed and enters airways.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Octadecylamine, 90%, tech., Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.