missing translation for 'onlineSavingsMsg'
Learn More

Octafluoronaphthalene, 96%, Thermo Scientific Chemicals

Product Code. 11400297
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Insoluble in water. Solubility in toluene, almost transparent.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 313-72-4
Molecular Formula C10F8
Molecular Weight (g/mol) 272.097
MDL Number MFCD00014307
InChI Key JDCMOHAFGDQQJX-UHFFFAOYSA-N
Synonym octafluoronaphthalene, perfluoronaphthalene, naphthalene, octafluoro, octafluoronapthalene, octafiuoronaphthalene, acmc-1cr7y, octafluoronaphthalene 1g, octafluoronaphthalene radical anion
PubChem CID 67564
IUPAC Name 1,2,3,4,5,6,7,8-octafluoronaphthalene
SMILES C12=C(C(=C(C(=C1F)F)F)F)C(=C(C(=C2F)F)F)F

Specifications

Melting Point 84°C to 89°C
Boiling Point 209°C
Quantity 5 g
Beilstein 1915630
Solubility Information Insoluble in water. Solubility in toluene,almost transparent.
Formula Weight 272.1
Percent Purity 96%
Chemical Name or Material Octafluoronaphthalene

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'