missing translation for 'onlineSavingsMsg'
Learn More

Octamethylcyclotetrasilazane, 97%, Thermo Scientific Chemicals

Chemical Identifiers

CAS 1020-84-4
Molecular Formula C8H28N4Si4
Molecular Weight (g/mol) 292.68
MDL Number MFCD00046123
InChI Key FIADVASZMLCQIF-UHFFFAOYSA-N
IUPAC Name 2,2,4,4,6,6,8,8-octamethyl-1,3,5,7,2,4,6,8-tetrazatetrasilocane
SMILES C[Si]1(C)N[Si](C)(C)N[Si](C)(C)N[Si](C)(C)N1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11339235
View Documents Promotion Details
Thermo Scientific Alfa Aesar
L16563.06
5 g N/A
N/A
11349235
View Documents Promotion Details
Thermo Scientific Alfa Aesar
L16563.14
25 g N/A
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1020-84-4
292.68
FIADVASZMLCQIF-UHFFFAOYSA-N
C[Si]1(C)N[Si](C)(C)N[Si](C)(C)N[Si](C)(C)N1
C8H28N4Si4
MFCD00046123
2,2,4,4,6,6,8,8-octamethyl-1,3,5,7,2,4,6,8-tetrazatetrasilocane

Specifications

224°C to 225°C
1020-84-4
MFCD00046123
976248
FIADVASZMLCQIF-UHFFFAOYSA-N
2,2,4,4,6,6,8,8-octamethyl-1,3,5,7,2,4,6,8-tetrazatetrasilocane
292.68
Ammonia-like
Octamethylcyclotetrasilazane
94°C to 98°C
C8H28N4Si4
UN2923
66°C (150°F)
C[Si]1(C)N[Si](C)(C)N[Si](C)(C)N[Si](C)(C)N1
292.68
97%
Moisture sensitive
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H314-H318
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H301-H314

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.

missing translation for 'einecsNumber' : 213-817-4

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only