missing translation for 'onlineSavingsMsg'
Learn More

Octane, ≥99% (GC), Honeywell™ Riedel-de-Haën™

Código de producto. 15607040
missing translation for 'orderingAttributeHoverText'
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
Este artículo no se puede devolver. Vea la política de devoluciones
Normativa: este producto está sujeto a impuestos especiales sobre productos energéticos de acuerdo con la Directiva Europea 2003/96 / CEE.
Este artículo no se puede devolver. Vea la política de devoluciones
Normativa: este producto está sujeto a impuestos especiales sobre productos energéticos de acuerdo con la Directiva Europea 2003/96 / CEE.

Puriss. p.a.,≥99.0% (GC)

  • Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.
TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 111-65-9
Molecular Formula C8H18
Molecular Weight (g/mol) 114.232
MDL Number MFCD00009556
InChI Key TVMXDCGIABBOFY-UHFFFAOYSA-N
Synonym n-octane, oktan, oktanen, ottani, oktanen dutch, heptane, methyl, oktan polish, ottani italian, octane, all isomers, unii-x1rv0b2fjv
PubChem CID 356
ChEBI CHEBI:17590
IUPAC Name octane
SMILES CCCCCCCC

Especificaciones

Melting Point -57°C
Density 0.703g/mL at 25°C
Boiling Point 125°C to 127°C
Flash Point 13°C (55.4°F)
Packaging Glass bottle
Linear Formula CH3(CH2)6CH3
Refractive Index 1.398
Quantity 500 mL
UN Number UN1262
Beilstein 1696875
Vapor Density 3.9 (vs air)
Formula Weight 114.23g/mol
Percent Purity ≥99.0% (GC)
Chemical Name or Material Octane
Mostrar más Mostrar menos
compliance-icons compliance-icons compliance-icons compliance-icons
Nombre del producto
  • Octane
Palabra de advertencia
  • Danger
Clasificación
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Aspiration toxicity Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Indicaciones de peligro
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Consejos de prudencia
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P243-Take precautionary measures against static discharge.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Otros peligros
  • MIXTURE LIST-Contains : octane
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'