missing translation for 'onlineSavingsMsg'
Learn More

Motic™ Widefield Eyepiece N-WF20X/13mm (ESD) for SMZ-171

Increases magnification to 20X. Motic™ Widefield Eyepiece N-WF20X/13mm (ESD) to your SMZ-171 to have more magnification (sold by 1 unit)

Brand:  Motic™ 1101001402741

Product Code. 15227169

  • 118.00 € / Each
Estimated Shipment: 11-06-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • For use with SMZ-171 Series Microscopes
  • Magnification: 20X
  • Field Number: 13mm
  • ESD safe

Order Info

dimProductType

Specifications

Specifications

Ocular Widefield Eyepiece
13 mm
20X
1 Ea.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Motic™ Widefield Eyepiece N-WF20X/13mm (ESD) for SMZ-171 > Product Type: Ocular; Magnification Power: 20X

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.