Learn More
Implen OD600™ IQOQ Package Documentation
IQ/OQ Documentation package for the Implen OD600 (provided as Excel on USB stick) including two Neutral Density Glass filters.
Brand:  Implen Implen OD600-Q
Product Code. 30093035
  • 1650.00 € / 1 set
Explore more special offers
Description
Description
  • IQOQ package for validation of the Implen OD600 instrument.
  • Includes
    • Two glass filters for validation of photometric range and accuracy.
    • IQOQ documentation provided as Excel file on USB stick
Specifications
Specifications
Implen OD600
IQ/OQ Documentation