missing translation for 'onlineSavingsMsg'
Learn More

Ohaus™ Heavy-Duty Solution Balance

Ideal for high-capacity weighings—offers 20kg capacity and 1g readability. OHAUS Heavy-Duty Solution Balance with sturdy construction enables balance to withstand harsh environments and constant use without sacrificing precision.

Brand:  Ohaus™ 80000000

Product Code. 10591022

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers

Includes: Storage rack containing one metric weight set (one each of 1kg, 5kg, 10kg; and two of 2kg)
This item is not returnable. View return policy

Description

Description

Standard Features

  • Cast-aluminum beam and base, 11 in. diameter (28cm) stainless-steel plate, covered steel self-aligning bearings, precision-ground knives
  • To provide precise, reproducible data, the balance also incorporates front and rear parallax-free reading; notched beams with center-indicating poise
  • Lockable tare for containers up to 2270g
  • Magnetic damping slows oscillation
  • Positive load stops prevent damage due to overloads
  • Corrosion-resistant coating on exposed parts ensures a long work life

Service Plan

Service agreement available.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

11 in.
11 in.
27.9 cm
28 cm
1119-D0
34 in.
44 lb., 20 kg
27.9 cm
100 x 1g (front beam), 1000 x 100g (rear beam)
11 in.
Storage rack containing one metric weight set
Stainless Steel
86.36 cm
Heavyduty Solution Balance
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers