missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OHAUS™ Triple Beam Balances
Accurate, rugged and dependable. OHAUS Triple Beam Balances have stainless-steel weighing pans and bearing covers which prevent rust and corrosion.
Brand: OHAUS™ 80000006
Alert:
CHOKING HAZARD (1). Small parts. Not for children under 3 yrs.
Description
- Precision-cast poises and deep notches in beams for accurate values every time
- Precision-ground knife edges and self-aligning agate bearings for maximum sensitivity and reproducibility
- Accurate with quick response time
- Magnetic damping minimizes oscillation
- Heavy zinc base won't tip easily like cast aluminum
Specifications
- Capacity: 610g (2610g with attachment weights)
- Readability: 0.1g
- Front Beam: 10 x 0.1g
- Middle Beam: 500 x 100g
- Rear Beam: 100 x 10g
- Vernier Dial (Dial-O-Gram™ Model only): 10 x 0.1g
- Pan size: 6 in. dia. (21.2cm)
- Dimensions: 18L x 12W x 9 in.H (45.7 x 30.5 x 22.3cm)
Dial-O-Gram model includes advanced features
- Direct-reading vernier dial for faster weighing results
- Tare beam allows containers up to 225g to be balanced out
- Add rod and clamp accessory (S40022B) to perform specific gravity determinations
Specific Gravity Rod and Clamp
- Suspends balance above base of table to measure specific gravity.
Poly Scoop and Counterweight
- Ideal for retaining all of the object being weighed
- Counterweight returns balance to zero when nothing but the scoop is on the platform
- Stainless-steel scoop and counterweight feature heavy-duty construction for long wear
Note
S40028 features a plastic container
Services
Five years
Specifications
6 in. | |
9 in. | |
22.86 cm | |
30.48 cm | |
Stainless Steel | |
18 in. | |
45.72 cm | |
1.34 lb., 0.61 kg |
15.2 cm | |
500 x 100g (middle beam), 100 x 10g (rear beam), 1 | |
12 in. | |
1650-00 | |
7.05 lb. | |
3.2 kg | |
5 Years | |
Triple Beam Dial-O-Gram™ Balance |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction